LIF372152787 Life Chemicals Inc.

2-mercapto-6,7-dihydro[1,3,4]thiadiazolo[3,2-{a}][1,3]diazepin-8(5{H})-one

  • MDL number MFCD04440711
  • Cas Number : 887201-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart