LIF372152814 Life Chemicals Inc.

2-(1H-Indol-3-ylsulfanyl)-ethylamine

  • MDL number MFCD00297155
  • Cas Number : 61021-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart