LIF372152819 Life Chemicals Inc.

[2-(2-Methyl-1H-indol-1-yl)ethyl]amine

  • MDL number MFCD04440779
  • Cas Number : 883535-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart