LIF372152820 Life Chemicals Inc.

4-Chloro-5,7-diphenyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD04971637
  • Cas Number : 287177-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart