LIF372152831 Life Chemicals Inc.

4-Chloro-5-phenyl-7-[3-(trifluoromethyl)phenyl]-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD03030413
  • Cas Number : 887202-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart