LIF372152832 Life Chemicals Inc.

5-phenyl-7-[3-(trifluoromethyl)phenyl]-7H-pyrrolo[2,3-d]pyrimidin-4-ol

  • MDL number MFCD04971643
  • Cas Number : 865546-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart