LIF372152838 Life Chemicals Inc.

4-(2,4,6-Trimethyl-phenyl)-thiazol-2-ylamine

  • MDL number MFCD01534593
  • Cas Number : 81529-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart