LIF372152842 Life Chemicals Inc.

4-(2-Amino-1,3-thiazol-4-yl)benzonitrile

  • MDL number MFCD03407828
  • Cas Number : 436151-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart