LIF372152845 Life Chemicals Inc.

Naphtho[2,1-d]thiazol-2-ylamine

  • MDL number MFCD00185989
  • Cas Number : 1203-55-0
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart