LIF372152849 Life Chemicals Inc.

1-(2-Isothiocyanatoethyl)-1H-indole

  • MDL number MFCD04440791
  • Cas Number : 887202-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart