LIF372152854 Life Chemicals Inc.

4-(4,6-Dichloro-pyrimidin-2-yl)-morpholine

  • MDL number MFCD05022359
  • Cas Number : 10397-13-4
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart