LIF372152857 Life Chemicals Inc.

6-(Piperidine-1-sulfonyl)-benzothiazol-2-ylamine

  • MDL number MFCD04441052
  • Cas Number : 854058-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart