LIF372152861 Life Chemicals Inc.

5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD03613706
  • Cas Number : 14466-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart