LIF372152863 Life Chemicals Inc.

1-benzyl-4-phenylpiperidin-4-ol

  • MDL number MFCD00598510
  • Cas Number : 63843-83-4
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart