LIF372152869 Life Chemicals Inc.

4-Amino-N-benzothiazol-2-yl-benzamide

  • MDL number MFCD00781842
  • Cas Number : 36855-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart