LIF372152876 Life Chemicals Inc.

N-(1-Adamantylmethyl)-4-(2-hydroxyethyl)piperazine-1-carboxamide

  • Cas Number : 612802-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart