LIF372152886 Life Chemicals Inc.

2-(3-methylphenoxy)acetic acid

  • MDL number MFCD00014358
  • Cas Number : 1643-15-8
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart