LIF372152896 Life Chemicals Inc.

ethyl 3-anilino-3-oxopropanoate

  • MDL number MFCD00956734
  • Cas Number : 53341-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart