LIF372152897 Life Chemicals Inc.

(5,6-Diphenyl-furo[2,3-d]pyrimidin-4-ylamino)-acetic acid

  • MDL number MFCD05156420
  • Cas Number : 701223-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart