LIF372152899 Life Chemicals Inc.

1-[5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid

  • MDL number MFCD00209248
  • Cas Number : 406476-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart