LIF372152908 Life Chemicals Inc.

2,3-diphenylquinoxaline-6-carboxylic acid

  • MDL number MFCD00555042
  • Cas Number : 32387-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart