LIF372152909 Life Chemicals Inc.

4-allyl-5-[(2-amino-1,3-thiazol-4-yl)methyl]-2,4-dihydro-3H-1,2,4-triazole-3-thione

  • MDL number MFCD05719126
  • Cas Number : 690643-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart