LIF372152910 Life Chemicals Inc.

3-[(2-amino-1,3-thiazol-4-yl)methyl]-4-methyl-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD05719131
  • Cas Number : 690642-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart