LIF372152925 Life Chemicals Inc.

5-[(2-amino-1,3-thiazol-4-yl)methyl]-N-methyl-1,3,4-thiadiazol-2-amine

  • MDL number MFCD05841195
  • Cas Number : 690642-84-3
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart