LIF372152929 Life Chemicals Inc.

1-(2-Furylmethyl)-4-hydroxy-6-methylpyridin-2(1H)-one

  • MDL number MFCD04075129
  • Cas Number : 638139-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart