LIF372152949 Life Chemicals Inc.

1-(2,6-Dimethyl-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD05096196
  • Cas Number : 63674-48-6
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart