LIF372152951 Life Chemicals Inc.

2-[(2-amino-1,3-thiazol-4-yl)methyl]-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD00612533
  • Cas Number : 91494-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart