LIF372152964 Life Chemicals Inc.

3-(Aminocarbonyl)-2,2,3-trimethylcyclopentanecarboxylic acid

  • MDL number MFCD00029147
  • Cas Number : 5333-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart