LIF372152967 Life Chemicals Inc.

5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00548507
  • Cas Number : 1140-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart