LIF372152972 Life Chemicals Inc.

4-Hydroxy-3-methyl-4,5,6,7-tetrahydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD04026789
  • Cas Number : 575462-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart