LIF372152985 Life Chemicals Inc.

(6-Bromo-benzothiazol-2-yl)-hydrazine

  • MDL number MFCD04448804
  • Cas Number : 37390-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart