LIF372152988 Life Chemicals Inc.

4,7-Dichloro-2-hydrazino-1,3-benzothiazole

  • MDL number MFCD04448807
  • Cas Number : 872696-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart