LIF372152994 Life Chemicals Inc.

7-Chloro-2-hydrazino-4-methoxy-1,3-benzothiazole

  • MDL number MFCD09743041
  • Cas Number : 941867-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart