LIF372153000 Life Chemicals Inc.

2-Chloro-4-ethoxy-1,3-benzothiazole

  • MDL number MFCD04971816
  • Cas Number : 890091-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart