LIF372153003 Life Chemicals Inc.

6-butyl-2-chloro-1,3-benzothiazole

  • MDL number MFCD04971820
  • Cas Number : 156246-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart