LIF372153004 Life Chemicals Inc.

2,4,5-Trichloro-1,3-benzothiazole

  • MDL number MFCD04971823
  • Cas Number : 898747-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart