LIF372153005 Life Chemicals Inc.

2,6-Dichloro-4-methyl-1,3-benzothiazole

  • MDL number MFCD04971826
  • Cas Number : 81561-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart