LIF372153015 Life Chemicals Inc.

6-chloro-4-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD02664350
  • Cas Number : 38338-21-5
  • Molecular Weight : 198.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart