LIF372153017 Life Chemicals Inc.

[1,3]Dioxolo[4',5':4,5]benzo[1,2-d]thiazol-6-ylamine

  • MDL number MFCD04448828
  • Cas Number : 50850-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart