LIF372153028 Life Chemicals Inc.

1-(3-Methylphenyl)pyrimidine-2,4,6(1H,3H,5H)-trione

  • MDL number MFCD01052227
  • Cas Number : 109493-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart