LIF372153029 Life Chemicals Inc.

1-p-Tolyl-pyrimidine-2,4,6-trione

  • MDL number MFCD00815752
  • Cas Number : 16348-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart