LIF372153030 Life Chemicals Inc.

1-(2-Methoxy-phenyl)-pyrimidine-2,4,6-trione

  • MDL number MFCD00552551
  • Cas Number : 16348-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart