LIF372153033 Life Chemicals Inc.

1-(3,4-Dimethylphenyl)pyrimidine-2,4,6(1H,3H,5H)-trione

  • MDL number MFCD02193532
  • Cas Number : 340225-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart