LIF372153039 Life Chemicals Inc.

1-(2,5-Dimethoxyphenyl)pyrimidine-2,4,6(1H,3H,5H)-trione

  • MDL number MFCD01241504
  • Cas Number : 352441-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart