LIF372153041 Life Chemicals Inc.

N~2~-(4-aminophenyl)-N~4~,N~4~,6-trimethylpyrimidine-2,4-diamine

  • MDL number MFCD22587331
  • Cas Number : 1395786-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart