LIF372153050 Life Chemicals Inc.

3-Benzyl-4-chloro-2-oxo-2,3-dihydro-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD03085539
  • Cas Number : 497082-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart