LIF372153079 Life Chemicals Inc.

5-(1H-Indol-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03618465
  • Cas Number : 874782-55-5
  • Molecular Weight : 230.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart