LIF372153082 Life Chemicals Inc.

4-(4-fluorophenyl)-5-(1{H}-indol-3-yl)-4{H}-1,2,4-triazole-3-thiol

  • MDL number MFCD04971905
  • Cas Number : 887206-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart