LIF372153085 Life Chemicals Inc.

4-{[4-(2-Hydroxyethyl)piperazin-1-yl]sulfonyl}benzoic acid

  • MDL number MFCD03723347
  • Cas Number : 620103-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart