LIF372153086 Life Chemicals Inc.

N-[4-(Morpholin-4-ylsulfonyl)benzoyl]glycine

  • MDL number MFCD04971909
  • Cas Number : 712344-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart